Product Details

SNP ID
rs104893636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176151875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTGCGCGGGGTCACGGACAAG[A/T]GCCAGGCGGCCCCGGCGGTCACTAC
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 1482 Intron NP_008829.3
XM_005246509.3 1482 Intron XP_005246566.1
XM_005246510.4 1482 Intron XP_005246567.1
XM_005246511.3 1482 Intron XP_005246568.1
XM_005246513.4 1482 Intron XP_005246570.1
XM_006712477.2 1482 Intron XP_006712540.1
XM_011511065.2 1482 Intron XP_011509367.1
XM_011511066.2 1482 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014621.2 1482 Missense Mutation GAG,GTG E81V NP_055436.2
XM_005246514.4 1482 Missense Mutation GAG,GTG E81V XP_005246571.1
Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details