Product Details

SNP ID
rs114952168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34998594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGTAGGCAGTCAACCATGACTT[G/T]TACAGCTCCTGTGGAGAGCTGCAGG
Phenotype
MIM: 612471
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGXT2 PubMed Links

Gene Details

Gene
AGXT2
Gene Name
alanine--glyoxylate aminotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306173.1 1963 Intron NP_001293102.1
NM_031900.3 1963 UTR 3 NP_114106.1
XM_005248337.2 1963 UTR 3 XP_005248394.1
XM_005248338.2 1963 UTR 3 XP_005248395.1
XM_017009748.1 1963 UTR 3 XP_016865237.1

View Full Product Details