Product Details

SNP ID
rs111807548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55330750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATAAAACAGAAAAATGGCAAGA[C/T]TAGAATAGTACATCAGGTAGATTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFRAL PubMed Links

Gene Details

Gene
GFRAL
Gene Name
GDNF family receptor alpha like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207410.2 Intron NP_997293.2

View Full Product Details