Product Details
- SNP ID
-
rs116111965
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:55331777 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCAATAATTGCACATATTTAAGA[C/G]AGCAATGCTTACGTGATGCAAATGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GFRAL
PubMed Links
Gene Details
- Gene
- GFRAL
- Gene Name
- GDNF family receptor alpha like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207410.2 |
171 |
Missense Mutation |
CAG,GAG |
Q29E |
NP_997293.2 |
View Full Product Details