Product Details

SNP ID
rs116804658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90405535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTAGGCTCAGATTATTGCTA[C/T]TCTGTATTCAGATCTTCATGTGTCT
Phenotype
MIM: 611232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN12 PubMed Links

Gene Details

Gene
CLDN12
Gene Name
claudin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185072.2 187 UTR 5 NP_001172001.1
NM_001185073.2 187 UTR 5 NP_001172002.1
NM_012129.4 187 UTR 5 NP_036261.1
Gene
LOC101409256
Gene Name
cell division cycle 42 pseudogene
There are no transcripts associated with this gene.

Gene
LOC102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516764.2 187 Intron XP_011515066.1
XM_017012889.1 187 Intron XP_016868378.1
XM_017012890.1 187 Intron XP_016868379.1
XM_017012891.1 187 Intron XP_016868380.1
XM_017012892.1 187 UTR 5 XP_016868381.1
XM_017012893.1 187 UTR 5 XP_016868382.1
XM_017012894.1 187 Intron XP_016868383.1

View Full Product Details