Product Details

SNP ID
rs115193923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68300835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGCTACAAGGGACCCACTGGTA[C/G]CCCTTTTAGATTCTATCAAAAGGAA
Phenotype
MIM: 605641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC7A6 PubMed Links

Gene Details

Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076785.2 1409 UTR 3 NP_001070253.1
NM_003983.5 1409 UTR 3 NP_003974.3
XM_011523433.1 1409 UTR 3 XP_011521735.1
XM_011523434.1 1409 UTR 3 XP_011521736.1
XM_011523438.1 1409 UTR 3 XP_011521740.1
XM_011523439.1 1409 UTR 3 XP_011521741.1
XM_017023851.1 1409 UTR 3 XP_016879340.1
Gene
SLC7A6OS
Gene Name
solute carrier family 7 member 6 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032178.2 1409 UTR 3 NP_115554.2
XM_011523372.2 1409 Intron XP_011521674.1

View Full Product Details