Product Details

SNP ID
rs116880958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35333878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGGGATTAAATAAATTACAGAA[C/T]ATTCATGCAATGGAATAGTATATAG
Phenotype
MIM: 194534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 4639 Intron NP_001295052.1
NM_006965.3 4639 UTR 3 NP_008896.2
XM_005258341.3 4639 UTR 3 XP_005258398.1

View Full Product Details