Product Details

SNP ID
rs113100394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:36840245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTCTGTGTTGCCTGCTATCCA[A/G]TGAAAACTATTGTTTCCTGTTTTTT
Phenotype
MIM: 616480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1328 PubMed Links

Gene Details

Gene
KIAA1328
Gene Name
KIAA1328
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322327.1 Intron NP_001309256.1
NM_020776.1 Intron NP_065827.1
XM_005258315.4 Intron XP_005258372.1
XM_005258317.3 Intron XP_005258374.1
XM_005258318.4 Intron XP_005258375.1
XM_011526099.2 Intron XP_011524401.1
XM_011526100.2 Intron XP_011524402.1
XM_017025876.1 Intron XP_016881365.1
XM_017025877.1 Intron XP_016881366.1
XM_017025878.1 Intron XP_016881367.1
XM_017025879.1 Intron XP_016881368.1
XM_017025880.1 Intron XP_016881369.1
XM_017025881.1 Intron XP_016881370.1

View Full Product Details