Product Details
- SNP ID
-
rs115431286
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31259511 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAACTGGCAGATGCAGGAAATCAT[C/G]CAAAGAAATTGAGAGGAAGAAAGAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB115
PubMed Links
Gene Details
- Gene
- DEFB115
- Gene Name
- defensin beta 115
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037730.1 |
146 |
Missense Mutation |
TCC,TGC |
S49C |
NP_001032819.1 |
View Full Product Details