Product Details

SNP ID
rs115568656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:84678530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGGAGAAATTTTCAATGCAAG[C/T]AGATGCATCCAATTTCTCCAATTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDFY3 PubMed Links

Gene Details

Gene
WDFY3
Gene Name
WD repeat and FYVE domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014991.4 Intron NP_055806.2
XM_005262858.4 Intron XP_005262915.1
XM_011531757.2 Intron XP_011530059.1
XM_011531759.2 Intron XP_011530061.1
XM_011531760.2 Intron XP_011530062.1
XM_011531761.2 Intron XP_011530063.1
XM_011531762.2 Intron XP_011530064.1
XM_011531763.2 Intron XP_011530065.1
XM_011531764.2 Intron XP_011530066.1
XM_011531765.2 Intron XP_011530067.1
XM_011531766.2 Intron XP_011530068.1
XM_011531767.2 Intron XP_011530069.1
XM_017007906.1 Intron XP_016863395.1
XM_017007907.1 Intron XP_016863396.1
XM_017007908.1 Intron XP_016863397.1
XM_017007909.1 Intron XP_016863398.1

View Full Product Details