Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135597.1 | Intron | NP_001129069.1 | |||
NM_001254943.1 | Intron | NP_001241872.1 | |||
NM_018084.4 | Intron | NP_060554.3 | |||
XM_005264418.4 | Intron | XP_005264475.1 | |||
XM_005264421.2 | Intron | XP_005264478.1 | |||
XM_005264426.2 | Intron | XP_005264483.1 | |||
XM_011532965.2 | Intron | XP_011531267.1 | |||
XM_011532966.2 | Intron | XP_011531268.1 | |||
XM_011532967.2 | Intron | XP_011531269.1 | |||
XM_011532968.2 | Intron | XP_011531270.1 | |||
XM_017004476.1 | Intron | XP_016859965.1 | |||
XM_017004477.1 | Intron | XP_016859966.1 | |||
XM_017004478.1 | Intron | XP_016859967.1 | |||
XM_017004479.1 | Intron | XP_016859968.1 | |||
XM_017004480.1 | Intron | XP_016859969.1 | |||
XM_017004481.1 | Intron | XP_016859970.1 |