Product Details

SNP ID
rs116717282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:72998089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTCATCTAGATTTTGAGAAGA[A/G]GGGGAAAAAGGAAGTTTGTCCTGTC
Phenotype
MIM: 613822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4R2 PubMed Links

Gene Details

Gene
PPP4R2
Gene Name
protein phosphatase 4 regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318025.1 300 Intron NP_001304954.1
NM_001318026.1 300 Intron NP_001304955.1
NM_001318027.1 300 Intron NP_001304956.1
NM_001318028.1 300 Missense Mutation AAG,AGG K16R NP_001304957.1
NM_174907.3 300 Intron NP_777567.1

View Full Product Details