Product Details

SNP ID
rs115750534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:169864175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCCAAGGGGCTCACAGCCCAG[A/G]GCCTTTGCAAAGAAGCAGGGCAGGC
Phenotype
MIM: 603122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK2 PubMed Links

Gene Details

Gene
DOCK2
Gene Name
dedicator of cytokinesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004946.2 3088 Intron NP_004937.1
XM_005265830.3 3088 Intron XP_005265887.1
XM_011534448.2 3088 Intron XP_011532750.1
XM_011534449.2 3088 Intron XP_011532751.1
XM_011534450.2 3088 Intron XP_011532752.1
XM_011534451.2 3088 Intron XP_011532753.1
XM_017009189.1 3088 Intron XP_016864678.1
XM_017009190.1 3088 Intron XP_016864679.1
Gene
FAM196B
Gene Name
family with sequence similarity 196 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129891.1 3088 UTR 3 NP_001123363.1
XM_011534415.2 3088 UTR 3 XP_011532717.1
XM_017008908.1 3088 Intron XP_016864397.1

View Full Product Details