Product Details
- SNP ID
-
rs111888052
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:58402735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGAACTTTTCTTGCAAGAGGTAC[A/G]GATGGGAGAACTTTGGTTATCTAGA
- Phenotype
-
MIM: 607384
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TIMM9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7153357] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TIMM9
- Gene Name
- translocase of inner mitochondrial membrane 9
There are no transcripts associated with this gene.
- Gene
- TOMM20L
- Gene Name
- translocase of outer mitochondrial membrane 20 like
View Full Product Details