Product Details

SNP ID
rs115469051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150895053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGTTTACTTAAGCTTTCTTCCA[C/T]GTCTTTTATAGAAAAATTAGTTTAT
Phenotype
MIM: 608212 MIM: 612429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRGM PubMed Links

Gene Details

Gene
IRGM
Gene Name
immunity related GTPase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145805.1 2607 Intron NP_001139277.1
XM_011537641.2 2607 Intron XP_011535943.1
Gene
ZNF300
Gene Name
zinc finger protein 300
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172831.1 2607 UTR 3 NP_001166302.1
NM_001172832.1 2607 UTR 3 NP_001166303.1
NM_052860.2 2607 UTR 3 NP_443092.1
XM_011537702.2 2607 UTR 3 XP_011536004.1

View Full Product Details