Product Details

SNP ID
rs116998798
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49630415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGCTAAGGGCTTGGGGTGGGC[A/G]GGAAATCGGGCTTGAAGACCCTCCT
Phenotype
MIM: 616288
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FMNL3 PubMed Links

Gene Details

Gene
FMNL3
Gene Name
formin like 3
There are no transcripts associated with this gene.

Gene
PRPF40B
Gene Name
pre-mRNA processing factor 40 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031698.2 Intron NP_001026868.2
NM_012272.2 Intron NP_036404.1
XM_006719324.3 Intron XP_006719387.1
XM_006719325.3 Intron XP_006719388.1
XM_011538135.2 Intron XP_011536437.1
XM_011538136.2 Intron XP_011536438.1
XM_011538137.2 Intron XP_011536439.1
XM_011538138.2 Intron XP_011536440.1
XM_011538139.2 Intron XP_011536441.1
XM_011538140.2 Intron XP_011536442.1
XM_011538141.1 Intron XP_011536443.1
XM_011538143.2 Intron XP_011536445.1
XM_011538144.2 Intron XP_011536446.1
XM_017019135.1 Intron XP_016874624.1
XM_017019136.1 Intron XP_016874625.1
XM_017019137.1 Intron XP_016874626.1
XM_017019138.1 Intron XP_016874627.1
XM_017019139.1 Intron XP_016874628.1

View Full Product Details