Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323599.1 | Intron | NP_001310528.1 | |||
NM_001323600.1 | Intron | NP_001310529.1 | |||
NM_003174.3 | Intron | NP_003165.2 | |||
NM_021738.2 | Intron | NP_068506.2 | |||
XM_005252570.2 | Intron | XP_005252627.1 | |||
XM_005252571.3 | Intron | XP_005252628.1 | |||
XM_005252573.2 | Intron | XP_005252630.1 | |||
XM_011519630.2 | Intron | XP_011517932.2 | |||
XM_011519631.2 | Intron | XP_011517933.2 | |||
XM_011519632.2 | Intron | XP_011517934.2 | |||
XM_011519633.2 | Intron | XP_011517935.2 | |||
XM_011519634.1 | Intron | XP_011517936.1 | |||
XM_011519635.1 | Intron | XP_011517937.1 | |||
XM_011519636.2 | Intron | XP_011517938.2 | |||
XM_011519637.2 | Intron | XP_011517939.2 | |||
XM_011519638.2 | Intron | XP_011517940.2 | |||
XM_017016575.1 | Intron | XP_016872064.1 | |||
XM_017016576.1 | Intron | XP_016872065.1 | |||
XM_017016577.1 | Intron | XP_016872066.1 | |||
XM_017016578.1 | Intron | XP_016872067.1 | |||
XM_017016579.1 | Intron | XP_016872068.1 | |||
XM_017016580.1 | Intron | XP_016872069.1 | |||
XM_017016581.1 | Intron | XP_016872070.1 | |||
XM_017016582.1 | Intron | XP_016872071.1 | |||
XM_017016583.1 | Intron | XP_016872072.1 |