Product Details

SNP ID
rs117626671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99334265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATGTTCTCAGGCTGCAAGCCA[C/T]ATAACATAGTGGTCATTTTCTTTAG
Phenotype
MIM: 607802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNNM1 PubMed Links

Gene Details

Gene
CNNM1
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020348.2 Intron NP_065081.2
XM_005269692.3 Intron XP_005269749.2
XM_011539629.1 Intron XP_011537931.1
XM_011539630.1 Intron XP_011537932.1
XM_011539631.2 Intron XP_011537933.1

View Full Product Details