Product Details

SNP ID
rs147188948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117283664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCTGTAGGCAAGCCCCTGTA[C/T]AGATCACGACCAATTTCTTTAACTG
Phenotype
MIM: 614235 MIM: 193001
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDZD8 PubMed Links

Gene Details

Gene
PDZD8
Gene Name
PDZ domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173791.4 3159 Silent Mutation CTA,CTG L1023L NP_776152.1
XM_005269518.3 3159 Silent Mutation CTA,CTG L982L XP_005269575.1
XM_011539265.1 3159 Intron XP_011537567.1
XM_011539266.2 3159 Intron XP_011537568.1
Gene
SLC18A2
Gene Name
solute carrier family 18 member A2
There are no transcripts associated with this gene.

View Full Product Details