Product Details

SNP ID
rs151112513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86960017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCATAGTGGCCGAGAAGACCAA[A/G]GAGCAGGCCAACGCCGTGAGCGAGG
Phenotype
MIM: 608925 MIM: 602998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADIRF PubMed Links

Gene Details

Gene
ADIRF
Gene Name
adipogenesis regulatory factor
There are no transcripts associated with this gene.

Gene
MMRN2
Gene Name
multimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024756.2 347 Intron NP_079032.2
XM_005270153.1 347 Intron XP_005270210.1
XM_006717970.3 347 Intron XP_006718033.1
Gene
SNCG
Gene Name
synuclein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003087.2 347 Silent Mutation AAA,AAG K60K NP_003078.2
XM_011540105.2 347 Missense Mutation AGA,GGA R78G XP_011538407.1

View Full Product Details