Product Details

SNP ID
rs117111740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62433767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCGCGGAAGGTATTCCTTTAAC[C/T]GCAGGTGTTTGTTGCCCTTGGCACA
Phenotype
MIM: 103390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHNAK PubMed Links

Gene Details

Gene
AHNAK
Gene Name
AHNAK nucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001620.2 867 Intron NP_001611.1
NM_024060.3 867 UTR 3 NP_076965.2
XM_005274240.1 867 Intron XP_005274297.1
XM_005274241.2 867 Intron XP_005274298.1
XM_005274245.1 867 Intron XP_005274302.1
XM_017018270.1 867 Intron XP_016873759.1
XM_017018271.1 867 Intron XP_016873760.1
XM_017018272.1 867 Intron XP_016873761.1
XM_017018273.1 867 Intron XP_016873762.1

View Full Product Details