Product Details

SNP ID
rs137985207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCACTGTGATGGCCTATGACC[A/G]CTACGTGGCCATCTGTAAGCCCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 359 Missense Mutation CAC,CGC H120R NP_001004704.1

View Full Product Details