Product Details

SNP ID
rs142444931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10576236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCATGACCTGATGGGATGGG[C/T]GGCAGTGTGTCCACACTTGGGCCTG
Phenotype
MIM: 605702 MIM: 604673
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYVE1 PubMed Links

Gene Details

Gene
LYVE1
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
There are no transcripts associated with this gene.

Gene
MRVI1
Gene Name
murine retrovirus integration site 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098579.2 3168 UTR 3 NP_001092049.2
NM_001100163.2 3168 UTR 3 NP_001093633.1
NM_001100167.2 3168 UTR 3 NP_001093637.1
NM_001206880.1 3168 UTR 3 NP_001193809.1
NM_001206881.1 3168 UTR 3 NP_001193810.1
NM_130385.3 3168 UTR 3 NP_569056.4
Gene
MRVI1-AS1
Gene Name
MRVI1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details