Product Details

SNP ID
rs144457255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66473291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTTCGTGGTAACAGACACGTC[A/C]CCTGGAGGAGGGGCTGCCGAGGGCC
Phenotype
MIM: 606818 MIM: 609827
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DPP3 PubMed Links

Gene Details

Gene
DPP3
Gene Name
dipeptidyl peptidase 3
There are no transcripts associated with this gene.

Gene
LOC101928069
Gene Name
uncharacterized LOC101928069
There are no transcripts associated with this gene.

Gene
PELI3
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098510.1 438 Silent Mutation TCA,TCC S145S NP_001091980.1
NM_001243135.1 438 Silent Mutation TCA,TCC S106S NP_001230064.1
NM_001243136.1 438 Silent Mutation TCA,TCC S62S NP_001230065.1
NM_145065.2 438 Silent Mutation TCA,TCC S169S NP_659502.2
XM_011544884.2 438 Silent Mutation TCA,TCC S169S XP_011543186.1
XM_017017465.1 438 Silent Mutation TCA,TCC S20S XP_016872954.1

View Full Product Details