Product Details

SNP ID
rs146965889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCACTGTGATGGCCTATGAC[C/T]GCTACGTGGCCATCTGTAAGCCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 358 Missense Mutation CGC,TGC R120C NP_001004704.1

View Full Product Details