Product Details

SNP ID
rs144111044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6935780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTTTCCTCCTTCCCCTCAACC[A/G]CCAGACAGCACCCCTCGACAGCCAG
Phenotype
MIM: 607462 MIM: 615140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007026.1 743 Silent Mutation CCA,CCG P171P NP_001007027.1
NM_001940.3 743 Silent Mutation CCA,CCG P171P NP_001931.2
Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
There are no transcripts associated with this gene.

Gene
RNU7-1
Gene Name
RNA, U7 small nuclear 1
There are no transcripts associated with this gene.

View Full Product Details