Product Details

SNP ID
rs138752044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20405625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTACCTTGCTGGTTTGGGTGGA[G/T]GGAGCCAGGCGCTCATCTGCACAAG
Phenotype
MIM: 609877
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYL1 PubMed Links

Gene Details

Gene
CRYL1
Gene Name
crystallin lambda 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015974.2 Intron NP_057058.2
XM_005266415.3 Intron XP_005266472.1
XM_005266416.4 Intron XP_005266473.1
XM_017020624.1 Intron XP_016876113.1

View Full Product Details