Product Details

SNP ID
rs149222191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27623182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAATGTCTGCCAACATGTGCTT[G/T]ACAAGTTTGAGGCCAGCATAAAGGA
Phenotype
MIM: 609733 MIM: 613715
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
There are no transcripts associated with this gene.

Gene
POLR1D
Gene Name
RNA polymerase I subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206559.1 1379 Intron NP_001193488.1
NM_015972.3 1379 Missense Mutation GAC,TAC D112Y NP_057056.1
NM_152705.2 1379 Intron NP_689918.1
XM_005266412.1 1379 Missense Mutation GAC,TAC D112Y XP_005266469.1
XM_005266414.2 1379 Intron XP_005266471.1
XM_017020622.1 1379 Intron XP_016876111.1
XM_017020623.1 1379 Intron XP_016876112.1

View Full Product Details