Product Details

SNP ID
rs142747516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60734864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTCTGTAGGAGGGAAACCGCCA[C/T]GGACGATCAGGGTTGCCCTCGGTGT
Phenotype
MIM: 602659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MNAT1 PubMed Links

Gene Details

Gene
MNAT1
Gene Name
MNAT1, CDK activating kinase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177963.1 130 Missense Mutation ACG,ATG T1M NP_001171434.1
NM_002431.3 130 Missense Mutation ACG,ATG T1M NP_002422.1
XM_005267687.2 130 Missense Mutation ACG,ATG T1M XP_005267744.1
XM_005267688.2 130 Missense Mutation ACG,ATG T1M XP_005267745.1
XM_017021332.1 130 Intron XP_016876821.1
XM_017021333.1 130 UTR 5 XP_016876822.1
XM_017021334.1 130 Intron XP_016876823.1

View Full Product Details