Product Details

SNP ID
rs139920166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68303658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGTTTCTCTACTTCTAAGGTAC[A/G]AATTAAAAAATAGATTTTGTTGTCA
Phenotype
MIM: 610087 MIM: 605641
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRMT7 PubMed Links

Gene Details

Gene
PRMT7
Gene Name
protein arginine methyltransferase 7
There are no transcripts associated with this gene.

Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
There are no transcripts associated with this gene.

Gene
SLC7A6OS
Gene Name
solute carrier family 7 member 6 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032178.2 Intron NP_115554.2
XM_011523372.2 Intron XP_011521674.1

View Full Product Details