Product Details

SNP ID
rs140343297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3090195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCTCAGGTGCGAGCTCTGGCG[A/G]AAGCGGTGGCCGCACAGGTGGCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 1166 Silent Mutation TTC,TTT F19F NP_001269344.1
NM_001282416.1 1166 Silent Mutation TTC,TTT F276F NP_001269345.1
NM_032805.2 1166 Silent Mutation TTC,TTT F413F NP_116194.2
XM_017023791.1 1166 Silent Mutation TTC,TTT F413F XP_016879280.1
XM_017023792.1 1166 Silent Mutation TTC,TTT F381F XP_016879281.1

View Full Product Details