Product Details

SNP ID
rs145295505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30108378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATCTTGCCCACCTCACCATTT[C/T]GAAACCACAGCCAATAACTGGTTCA
Phenotype
MIM: 616318 MIM: 601795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDPD3 PubMed Links

Gene Details

Gene
GDPD3
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024307.2 841 Silent Mutation TCA,TCG S254S NP_077283.2
Gene
LOC101928595
Gene Name
uncharacterized LOC101928595
There are no transcripts associated with this gene.

Gene
MAPK3
Gene Name
mitogen-activated protein kinase 3
There are no transcripts associated with this gene.

View Full Product Details