Product Details

SNP ID
rs146684809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31076469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGACAGCATCCCTCGGCCCTAC[C/T]GTTGTCAGCAGTGTGGGCGGGGCTA
Phenotype
MIM: 610561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 1213 Missense Mutation CGT,TGT R49C NP_055514.3
XM_005255710.3 1213 Missense Mutation CGT,TGT R49C XP_005255767.1
XM_005255711.3 1213 Missense Mutation CGT,TGT R49C XP_005255768.1
XM_005255712.3 1213 Missense Mutation CGT,TGT R49C XP_005255769.1
XM_011545990.2 1213 Missense Mutation CGT,TGT R49C XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
There are no transcripts associated with this gene.

View Full Product Details