Product Details

SNP ID
rs149566187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4959878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGACACTAAGTTCTGTATCTA[C/T]GCAGAGGTGGAGTCGGGGCTGAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC14L5 PubMed Links

Gene Details

Gene
SEC14L5
Gene Name
SEC14 like lipid binding 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014692.1 Intron NP_055507.1

View Full Product Details