Product Details

SNP ID
rs149626751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19725516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCTGGGCCCCTGATTTTCTTA[A/C]GTATTTCTAATGCAGAACAGAGCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IQCK PubMed Links

Gene Details

Gene
IQCK
Gene Name
IQ motif containing K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305121.1 Intron NP_001292050.1
NM_001305122.1 Intron NP_001292051.1
NM_153208.2 Intron NP_694940.1
XM_011545736.2 Intron XP_011544038.1
XM_017022933.1 Intron XP_016878422.1
Gene
KNOP1
Gene Name
lysine rich nucleolar protein 1
There are no transcripts associated with this gene.

View Full Product Details