Product Details

SNP ID
rs141941876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35918417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATACCCTTCCTCCTCTTGGCC[A/G]TACTGCTTCCAGGGAGAGCAAGGGA
Phenotype
MIM: 612896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A8P PubMed Links

Gene Details

Gene
LRRC37A8P
Gene Name
leucine rich repeat containing 37 member A8, pseudogene
There are no transcripts associated with this gene.

Gene
RDM1
Gene Name
RAD52 motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034836.1 730 Intron NP_001030008.1
NM_001163120.1 730 Silent Mutation TAC,TAT Y142Y NP_001156592.1
NM_001163121.1 730 Silent Mutation TAC,TAT Y227Y NP_001156593.1
NM_001163122.1 730 Silent Mutation TAC,TAT Y204Y NP_001156594.1
NM_001163124.1 730 Silent Mutation TAC,TAT Y119Y NP_001156596.1
NM_001163125.1 730 Intron NP_001156597.1
NM_001163130.1 730 Silent Mutation TAC,TAT Y237Y NP_001156602.1
NM_145654.3 730 Silent Mutation TAC,TAT Y260Y NP_663629.1
XM_011524509.2 730 Intron XP_011522811.1
XM_011524510.2 730 Intron XP_011522812.1

View Full Product Details