Product Details

SNP ID
rs143769514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36545793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTTCGAGAGGAGGCAGATTCC[C/G]CTCTACATAGACGACACCCTGACGG
Phenotype
MIM: 612275 MIM: 610275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GGNBP2 PubMed Links

Gene Details

Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024835.4 361 Silent Mutation CCC,CCG P23P NP_079111.1
XM_005257689.2 361 Silent Mutation CCC,CCG P23P XP_005257746.1
XM_011525265.2 361 Intron XP_011523567.1
XM_011525266.1 361 Silent Mutation CCC,CCG P23P XP_011523568.1
XM_017025107.1 361 Intron XP_016880596.1
Gene
PIGW
Gene Name
phosphatidylinositol glycan anchor biosynthesis class W
There are no transcripts associated with this gene.

View Full Product Details