Product Details
- SNP ID
-
rs149597713
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8803351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCTTTGATTGCTTCCACCTCCTG[C/T]TGCCCATGCAAATTGAGCCACGGTG
- Phenotype
-
MIM: 611462
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MFSD6L
PubMed Links
Gene Details
- Gene
- MFSD6L
- Gene Name
- major facilitator superfamily domain containing 6 like
There are no transcripts associated with this gene.
- Gene
- PIK3R6
- Gene Name
- phosphoinositide-3-kinase regulatory subunit 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010855.3 |
2009 |
Silent Mutation |
CAA,CAG |
Q729Q |
NP_001010855.1 |
NM_001290211.1 |
2009 |
Silent Mutation |
CAA,CAG |
Q593Q |
NP_001277140.1 |
XM_011523677.2 |
2009 |
Missense Mutation |
ACA,GCA |
T690A |
XP_011521979.1 |
XM_011523678.2 |
2009 |
Missense Mutation |
ACA,GCA |
T686A |
XP_011521980.1 |
XM_011523680.2 |
2009 |
Missense Mutation |
ACA,GCA |
T650A |
XP_011521982.1 |
XM_011523681.2 |
2009 |
Missense Mutation |
ACA,GCA |
T611A |
XP_011521983.1 |
XM_011523683.2 |
2009 |
Missense Mutation |
ACA,GCA |
T611A |
XP_011521985.1 |
XM_011523686.2 |
2009 |
Missense Mutation |
ACA,GCA |
T402A |
XP_011521988.1 |
XM_017024228.1 |
2009 |
Missense Mutation |
ACA,GCA |
T691A |
XP_016879717.1 |
XM_017024229.1 |
2009 |
Silent Mutation |
CAA,CAG |
Q729Q |
XP_016879718.1 |
XM_017024230.1 |
2009 |
Silent Mutation |
CAA,CAG |
Q688Q |
XP_016879719.1 |
XM_017024231.1 |
2009 |
Missense Mutation |
ACA,GCA |
T611A |
XP_016879720.1 |
View Full Product Details