Product Details

SNP ID
rs137996950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39532435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCTTCCCTCGCCTTGCAGCCT[A/T]CCACAAACAGCCTGCGGCGTTCCTC
Phenotype
MIM: 609773
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EID2 PubMed Links

Gene Details

Gene
EID2
Gene Name
EP300 interacting inhibitor of differentiation 2
There are no transcripts associated with this gene.

Gene
EID2B
Gene Name
EP300 interacting inhibitor of differentiation 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152361.2 433 Missense Mutation GAA,GTA E123V NP_689574.1

View Full Product Details