Product Details

SNP ID
rs139442151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49645047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCATGGAGGAAGAAGATGAGTC[A/T]CGAGGGAAGACAGAGGAGTCGGGGG
Phenotype
MIM: 165090
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RRAS PubMed Links

Gene Details

Gene
RRAS
Gene Name
related RAS viral (r-ras) oncogene homolog
There are no transcripts associated with this gene.

Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021228.2 70 Silent Mutation TCA,TCT S7S NP_067051.2
XM_005259122.4 70 Silent Mutation TCA,TCT S7S XP_005259179.1
XM_011527194.2 70 Silent Mutation TCA,TCT S10S XP_011525496.1
XM_017027083.1 70 Intron XP_016882572.1

View Full Product Details