Product Details
- SNP ID
-
rs140496718
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55181894 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAGGATGCACTGATGCAGGAATA[C/T]GTACTGAGCCTGGGAAGCAGGCACG
- Phenotype
-
MIM: 602510
MIM: 600782
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PTPRH
PubMed Links
Gene Details
- Gene
- PTPRH
- Gene Name
- protein tyrosine phosphatase, receptor type H
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161440.2 |
3302 |
Missense Mutation |
ATA,GTA |
I892V |
NP_001154912.2 |
NM_002842.4 |
3302 |
Missense Mutation |
ATA,GTA |
I1070V |
NP_002833.4 |
XM_011527183.1 |
3302 |
Missense Mutation |
ATA,GTA |
I1077V |
XP_011525485.1 |
XM_011527188.1 |
3302 |
Missense Mutation |
ATA,GTA |
I883V |
XP_011525490.1 |
XM_011527190.1 |
3302 |
Intron |
|
|
XP_011525492.1 |
XM_017027056.1 |
3302 |
Missense Mutation |
ATA,GTA |
I1092V |
XP_016882545.1 |
XM_017027057.1 |
3302 |
Missense Mutation |
ATA,GTA |
I1088V |
XP_016882546.1 |
XM_017027058.1 |
3302 |
Missense Mutation |
ATA,GTA |
I1085V |
XP_016882547.1 |
XM_017027059.1 |
3302 |
Intron |
|
|
XP_016882548.1 |
XM_017027060.1 |
3302 |
Missense Mutation |
ATA,GTA |
I1076V |
XP_016882549.1 |
XM_017027061.1 |
3302 |
Missense Mutation |
ATA,GTA |
I914V |
XP_016882550.1 |
XM_017027062.1 |
3302 |
Missense Mutation |
ATA,GTA |
I883V |
XP_016882551.1 |
XM_017027063.1 |
3302 |
Missense Mutation |
ATA,GTA |
I883V |
XP_016882552.1 |
XM_017027064.1 |
3302 |
Intron |
|
|
XP_016882553.1 |
- Gene
- SYT5
- Gene Name
- synaptotagmin 5
There are no transcripts associated with this gene.
View Full Product Details