Product Details

SNP ID
rs142978273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45408754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCCCAAAGGGAAAGAAACCTT[C/T]GAGCCAGAAGACAAGACAGTGAAGC
Phenotype
MIM: 107325 MIM: 126380 MIM: 607463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD3EAP PubMed Links

Gene Details

Gene
CD3EAP
Gene Name
CD3e molecule associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297590.1 1889 Silent Mutation TTC,TTT F264F NP_001284519.1
NM_012099.1 1889 Silent Mutation TTC,TTT F262F NP_036231.1
XM_017026180.1 1889 Silent Mutation TTC,TTT F84F XP_016881669.1
XM_017026181.1 1889 Silent Mutation TTC,TTT F84F XP_016881670.1
Gene
ERCC1
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166049.1 1889 UTR 3 NP_001159521.1
NM_001983.3 1889 UTR 3 NP_001974.1
NM_202001.2 1889 Intron NP_973730.1
XM_005258634.1 1889 Intron XP_005258691.1
XM_005258635.2 1889 Intron XP_005258692.1
XM_005258636.4 1889 Intron XP_005258693.1
XM_005258637.1 1889 Intron XP_005258694.1
XM_011526610.2 1889 Intron XP_011524912.1
XM_017026459.1 1889 Intron XP_016881948.1
XM_017026460.1 1889 UTR 3 XP_016881949.1
XM_017026461.1 1889 UTR 3 XP_016881950.1
XM_017026462.1 1889 UTR 3 XP_016881951.1
XM_017026463.1 1889 UTR 3 XP_016881952.1
XM_017026464.1 1889 UTR 3 XP_016881953.1
XM_017026465.1 1889 UTR 3 XP_016881954.1
XM_017026466.1 1889 UTR 3 XP_016881955.1
Gene
PPP1R13L
Gene Name
protein phosphatase 1 regulatory subunit 13 like
There are no transcripts associated with this gene.

View Full Product Details