Product Details

SNP ID
rs147281464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46469819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCCTCGGCAGGCTGCTGGCCC[C/T]GAGGGGCCTTCTGATCTGATGCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNMAL1 PubMed Links

Gene Details

Gene
PNMAL1
Gene Name
paraneoplastic Ma antigen family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103149.1 1537 Intron NP_001096619.1
NM_018215.3 1537 Missense Mutation CAG,CGG Q406R NP_060685.2
XM_011527067.2 1537 Missense Mutation CAG,CGG Q406R XP_011525369.1

View Full Product Details