Product Details

SNP ID
rs111770759
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:235662964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCCGGCTGCATAGCTGCTAAG[A/G]AAGGAATAGAACATTGGCTGTTTCA
Phenotype
MIM: 606897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYST PubMed Links

Gene Details

Gene
LYST
Gene Name
lysosomal trafficking regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000081.3 12001 Silent Mutation TTC,TTT F3794F NP_000072.2
NM_001301365.1 12001 Silent Mutation TTC,TTT F3794F NP_001288294.1
XM_011544031.1 12001 Silent Mutation TTC,TTT F3848F XP_011542333.1
XM_011544032.1 12001 Silent Mutation TTC,TTT F3848F XP_011542334.1
XM_011544033.2 12001 Silent Mutation TTC,TTT F3848F XP_011542335.1
XM_011544034.1 12001 Silent Mutation TTC,TTT F3802F XP_011542336.1
XM_011544035.2 12001 Intron XP_011542337.1
XM_011544036.2 12001 Silent Mutation TTC,TTT F3069F XP_011542338.1
XM_011544037.2 12001 Intron XP_011542339.1
XM_011544039.2 12001 Intron XP_011542341.1
XM_011544040.2 12001 Intron XP_011542342.1
XM_017000150.1 12001 Silent Mutation TTC,TTT F3771F XP_016855639.1
XM_017000151.1 12001 Intron XP_016855640.1

View Full Product Details