Product Details

SNP ID
rs112725574
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:23030555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGAGAGAAATGCCAAAGCAGA[A/G]AAGGAAAAGAAGCTTCCCCCACCAC
Phenotype
MIM: 609132
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009999.2 592 Silent Mutation GAA,GAG E146E NP_001009999.1
NM_015013.3 592 Silent Mutation GAA,GAG E146E NP_055828.2
XM_005245786.2 592 Silent Mutation GAA,GAG E146E XP_005245843.1
XM_006710472.3 592 Silent Mutation GAA,GAG E146E XP_006710535.1
XM_006710473.2 592 Silent Mutation GAA,GAG E146E XP_006710536.1
XM_006710474.3 592 Silent Mutation GAA,GAG E146E XP_006710537.1
XM_017000716.1 592 Silent Mutation GAA,GAG E146E XP_016856205.1
XM_017000717.1 592 Silent Mutation GAA,GAG E146E XP_016856206.1
XM_017000718.1 592 UTR 5 XP_016856207.1
XM_017000719.1 592 UTR 5 XP_016856208.1
XM_017000720.1 592 UTR 5 XP_016856209.1

View Full Product Details