Product Details

SNP ID
rs114191610
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:77706743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGGTTTGGTACTATCTTACAC[A/G]ATGTAGTATATGCTCCAAATTAGAA
Phenotype
MIM: 615146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP33 PubMed Links

Gene Details

Gene
USP33
Gene Name
ubiquitin specific peptidase 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015017.4 Intron NP_055832.3
NM_201624.2 Intron NP_963918.1
NM_201626.2 Intron NP_963920.1
XM_005270648.1 Intron XP_005270705.1
XM_005270649.1 Intron XP_005270706.1
XM_011541055.1 Intron XP_011539357.1
XM_011541056.2 Intron XP_011539358.1
XM_017000722.1 Intron XP_016856211.1
XM_017000723.1 Intron XP_016856212.1
XM_017000724.1 Intron XP_016856213.1
XM_017000725.1 Intron XP_016856214.1
XM_017000726.1 Intron XP_016856215.1
XM_017000727.1 Intron XP_016856216.1
XM_017000728.1 Intron XP_016856217.1
XM_017000729.1 Intron XP_016856218.1

View Full Product Details