Product Details

SNP ID
rs116830281
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:235661441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGGAGTAATCTCATCTCTGCA[A/C]CTCCATGGACCTCAGGATTCACACC
Phenotype
MIM: 606897
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYST PubMed Links

Gene Details

Gene
LYST
Gene Name
lysosomal trafficking regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000081.3 13524 UTR 3 NP_000072.2
NM_001301365.1 13524 UTR 3 NP_001288294.1
XM_011544031.1 13524 UTR 3 XP_011542333.1
XM_011544032.1 13524 UTR 3 XP_011542334.1
XM_011544033.2 13524 UTR 3 XP_011542335.1
XM_011544034.1 13524 UTR 3 XP_011542336.1
XM_011544035.2 13524 Intron XP_011542337.1
XM_011544036.2 13524 UTR 3 XP_011542338.1
XM_011544037.2 13524 Intron XP_011542339.1
XM_011544039.2 13524 Intron XP_011542341.1
XM_011544040.2 13524 Intron XP_011542342.1
XM_017000150.1 13524 UTR 3 XP_016855639.1
XM_017000151.1 13524 Intron XP_016855640.1

View Full Product Details