Product Details

SNP ID
rs138285095
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:206931778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCTGACATGCTAATGTCTGTC[C/T]TGTAGCTTCTGATTGAAACTCGGTC
Phenotype
MIM: 606015 MIM: 173880
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 2217 Missense Mutation AAG,AGG K678R NP_002635.2
XM_011509629.1 2217 Missense Mutation AAG,AGG K678R XP_011507931.1

View Full Product Details