Product Details

SNP ID
rs139047952
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:158398691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGACTTGGACTTGGGCCGCAGA[C/T]AGATGATAGAGGCACAGCCATAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 776 Missense Mutation TCT,TTT S259F NP_001004475.1

View Full Product Details