Product Details

SNP ID
rs139967751
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:222722488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGCTTTATGGTATACCAAATA[C/T]GCTTCAAGACTGGCTGGAAAAGAAA
Phenotype
MIM: 612375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
There are no transcripts associated with this gene.

Gene
BROX
Gene Name
BRO1 domain and CAAX motif containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288579.1 850 Intron NP_001275508.1
NM_001288580.1 850 Intron NP_001275509.1
NM_001288581.1 850 Silent Mutation TAC,TAT Y125Y NP_001275510.1
NM_144695.3 850 Silent Mutation TAC,TAT Y125Y NP_653296.2
XM_005273065.2 850 Silent Mutation TAC,TAT Y125Y XP_005273122.2
XM_005273069.4 850 Silent Mutation TAC,TAT Y70Y XP_005273126.1
XM_006711173.3 850 Silent Mutation TAC,TAT Y142Y XP_006711236.1
XM_011509212.2 850 Intron XP_011507514.1
XM_011509213.2 850 Silent Mutation TAC,TAT Y139Y XP_011507515.1
XM_011509214.2 850 Silent Mutation TAC,TAT Y125Y XP_011507516.1
XM_017000374.1 850 Silent Mutation TAC,TAT Y125Y XP_016855863.1
XM_017000375.1 850 Intron XP_016855864.1
XM_017000376.1 850 Silent Mutation TAC,TAT Y70Y XP_016855865.1

View Full Product Details